5-Aminobenzene-1,2,3-tricarboxylic acid ≥96%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 5-Amino-1,2,3-benzenetricarboxylic acid, 1-Aminobenzene-3,4,5-tricarboxylic acid

A184082-1G A184082-25G A184082-5G
ALA184082-1GEA 60.71 USD
ALA184082-1G ALA184082-25G ALA184082-5G
5-Aminobenzene-1,2,3-tricarboxylic acid ≥96%
5-Aminobenzene-1,2,3-tricarboxylic acid
Formula: C₉H₇NO₆
Storage Temperature: Refrigerator
MDL Number: MFCD03839869
CAS Number: 37141-01-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4813a6cfb02d2f1e5a95ad1bdef4a92e21395ea&vid=e4813a6cfb02d2f1e5a95ad1bdef4a92e21395ea&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR