7-Amino-3-Cephem-4-Carboxylic Acid ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 7-Anca, (6R,7R)-7-Amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-Amino-3-cephem-4-carboxylic acid

A184066-1G A184066-25G A184066-5G
ALA184066-1GEA 16.06 USD
ALA184066-1G ALA184066-25G ALA184066-5G
7-Amino-3-Cephem-4-Carboxylic Acid ≥98%
7-Amino-3-Cephem-4-Carboxylic Acid
Formula: C₇H₈N₂O₃S
MW: 200.21 g/mol
Melting Pt: 211-213°C
Storage Temperature: Freezer
MDL Number: MFCD08063340
CAS Number: 36923-17-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26e77bd77195d2d0242357b2b1e241b495fa5159&vid=26e77bd77195d2d0242357b2b1e241b495fa5159&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR