4-Amino-3-bromoquinoline ≥95%
Supplier: ALADDIN SCIENTIFIC
A184061-1G
A184061-5G
ALA184061-1GEA
26.51
USD
ALA184061-1G
ALA184061-5G
4-Amino-3-bromoquinoline ≥95%
4-Amino-3-bromoquinoline
Formula:
C₉H₇BrN₂ Storage Temperature: Ambient |
MDL Number:
MFCD00234502 CAS Number: 36825-36-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...