2-Amino-4-chloro-6-methylpyridine ≥95%
Supplier: ALADDIN SCIENTIFIC
Warning
|
Synonyms:
2-Amino-4-chloro-6-picoline, 4-Chloro-6-methylpyridin-2-amine
A184035-1G
A184035-250MG
A184035-50MG
A184035-5G
ALA184035-1GEA
112.96
USD
ALA184035-1G
ALA184035250MG
ALA184035-50MG
ALA184035-5G
2-Amino-4-chloro-6-methylpyridine ≥95%
2-Amino-4-chloro-6-methylpyridine
Formula:
C₆H₇ClN₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00800671 CAS Number: 36340-61-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...