3-(Aminomethyl)phenylboronic acid hydrochloride ≥95% (contains varying amounts of Anhydride)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: (3-(Aminomethyl)phenyl)boronic acid hydrochloride, 3-(Aminomethyl)benzeneboronic acid hydrochloride

A183949-1G A183949-250MG A183949-25G A183949-5G
ALA183949-1GEA 121.51 USD
ALA183949-1G ALA183949250MG ALA183949-25G ALA183949-5G
3-(Aminomethyl)phenylboronic acid hydrochloride ≥95% (contains varying amounts of Anhydride)
3-(Aminomethyl)phenylboronic acid hydrochloride
Formula: C₇H₁₁BClNO₂
MW: 187.43 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02093050
CAS Number: 352525-94-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79a775e0e831702baf5e474b3bccb194dc272bc0&vid=79a775e0e831702baf5e474b3bccb194dc272bc0&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR