3-Amino-2-bromo-5-methylpyridine ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 3-Amino-2-bromo-5-picoline, 5-Amino-6-bromo-3-picoline, 2-Bromo-5-methyl-3-pyridinamine

A183879-1G A183879-25G A183879-5G
ALA183879-1GEA 62.61 USD
ALA183879-1G ALA183879-25G ALA183879-5G
3-Amino-2-bromo-5-methylpyridine ≥98%
3-Amino-2-bromo-5-methylpyridine
Formula: C₆H₇BrN₂
MW: 187.04 g/mol
Melting Pt: 103-106°C
MDL Number: MFCD08277281
CAS Number: 34552-14-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9280e0c3d1f96b23e54dfd805d3ce6504bf28df3&vid=9280e0c3d1f96b23e54dfd805d3ce6504bf28df3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR