[3-(Aminomethyl)phenyl]methanol ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 3-(Aminomethyl)benzyl Alcohol

A183846-1G A183846-250MG A183846-25G A183846-5G
ALA183846-1GEA 113.91 USD
ALA183846-1G ALA183846250MG ALA183846-25G ALA183846-5G
[3-(Aminomethyl)phenyl]methanol ≥97%
[3-(Aminomethyl)phenyl]methanol
Formula: C₈H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD06212981
CAS Number: 34231-22-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=269aba8a3feca929c6ba7d51dca8e44b3a360d9a&vid=269aba8a3feca929c6ba7d51dca8e44b3a360d9a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR