5-Amino-6-chloroquinoline ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION

A183833-1G A183833-250MG A183833-5G
ALA183833-1GEA 72.11 USD
ALA183833-1G ALA183833250MG ALA183833-5G
5-Amino-6-chloroquinoline ≥97%
5-Amino-6-chloroquinoline
Formula: C₉H₇ClN₂
MW: 178.6200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02947117
CAS Number: 341010-40-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=169a2c44f8b62826637af3fa057c189388131682&vid=169a2c44f8b62826637af3fa057c189388131682&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR