4-Aminoisophthalic acid ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-Amino-1,3-benzenedicarboxylic acid: 4-Aminoisophthalic acid, 5-Carboxyanthranilic acid, 4-Aminobenzene-1,3-dicarboxylic acid

A183824-1G A183824-250MG A183824-5G
ALA183824-1GEA 327.66 USD
ALA183824-1G ALA183824250MG ALA183824-5G
4-Aminoisophthalic acid ≥97%
4-Aminoisophthalic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00102008
CAS Number: 33890-03-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58610ed7c480590c897e3c75f09ca487329e3229&vid=58610ed7c480590c897e3c75f09ca487329e3229&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR