N-(4-Fluoro-2-methylphenyl)acetamide ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Acetamido-5-fluorotoluene

A183713-1G A183713-25G A183713-5G
ALA183713-1GEA 32.21 USD
ALA183713-1G ALA183713-25G ALA183713-5G
N-(4-Fluoro-2-methylphenyl)acetamide ≥98%
N-(4-Fluoro-2-methylphenyl)acetamide
Formula: C₉H₁₀FNO
MW: 167.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861997
CAS Number: 326-65-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51a030c737c77228dee4d227355dbfacbb20f834&vid=51a030c737c77228dee4d227355dbfacbb20f834&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR