Ethyl-7-aminoheptanoate hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Ethyl 7-aminoheptanoate hydrochloride, 7-Amino-heptanoic acid ethyl ester hydrochloride

A183507-100G A183507-10G A183507-1G A183507-25G A183507-5G
ALA183507-100GEA 268.76 USD
ALA183507-100G ALA183507-10G ALA183507-1G ALA183507-25G ALA183507-5G
Ethyl-7-aminoheptanoate hydrochloride ≥98%
Ethyl-7-aminoheptanoate hydrochloride
Formula: C₉H₂₀ClNO₂
MW: 209.7100 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09743977
CAS Number: 29840-65-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44f50156393e573ce1f6f3b1c557850d93fc14ae&vid=44f50156393e573ce1f6f3b1c557850d93fc14ae&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR