6'-(Diethylamino)-3'-methyl-2'-(phenylamino)-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 7-Anilino-3-diethylamino-6-methyl fluoran

A183491-100G A183491-1G A183491-25G A183491-5G
ALA183491-100GEA 93.01 USD
ALA183491-100G ALA183491-1G ALA183491-25G ALA183491-5G
6'-(Diethylamino)-3'-methyl-2'-(phenylamino)-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one ≥98%
6'-(Diethylamino)-3'-methyl-2'-(phenylamino)-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one
Formula: C₃₁H₂₈N₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00603523
CAS Number: 29512-49-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68d120cf4798fa18ed2cf9d006c2b375ce7094bc&vid=68d120cf4798fa18ed2cf9d006c2b375ce7094bc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR