3-Aminobutan-1-ol ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Danger
|
Synonyms:
3-Amino-butan-1-ol
A183438-25G
A183438-5G
A183438-100G
A183438-1G
ALA183438-25GEA
67.36
USD
ALA183438-25G
ALA183438-5G
ALA183438-100G
ALA183438-1G
3-Aminobutan-1-ol ≥98%
3-Aminobutan-1-ol
Formula:
C₄H₁₁NO MW: 89.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03412694 CAS Number: 2867-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...