2-Amino-4,5-dichlorophenol ≥97%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
4,5-Dichloro-2-hydroxyaniline
A183425-1G
A183425-250MG
ALA183425-1GEA
78.76
USD
ALA183425-1G
ALA183425250MG
2-Amino-4,5-dichlorophenol ≥97%
2-Amino-4,5-dichlorophenol
Formula:
C₆H₅Cl₂NO MW: 178.016 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12406838 CAS Number: 28443-57-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...