N1,N1-Dimethylbenzene-1,2-diamine ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

A183422-100G A183422-1G A183422-250MG A183422-25G A183422-5G
ALA183422-100GEA 3262.21 USD
ALA183422-100G ALA183422-1G ALA183422250MG ALA183422-25G ALA183422-5G
N1,N1-Dimethylbenzene-1,2-diamine ≥97%
N1,N1-Dimethylbenzene-1,2-diamine
Formula: C₈H₁₂N₂
Storage Temperature: Ambient
MDL Number: MFCD01706706
CAS Number: 2836-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR