3-Amino-4-methylphenol ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 3-Amino-p-cresol, 5-Hydroxy-2-methylaniline

A183421-1G A183421-250MG A183421-25G A183421-5G
ALA183421-1GEA 55.01 USD
ALA183421-1G ALA183421250MG ALA183421-25G ALA183421-5G
3-Amino-4-methylphenol ≥98%
3-Amino-4-methylphenol
Formula: C₇H₉NO
MW: 123.16 g/mol
Melting Pt: 161 °C
Storage Temperature: Ambient
MDL Number: MFCD00075083
CAS Number: 2836-00-2
UN: 2512
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20ed3994c49a34e080f1c4228f234abfe7beef86&vid=20ed3994c49a34e080f1c4228f234abfe7beef86&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR