6-Acetamidopicolinic acid ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A183313-1G A183313-250MG A183313-25G A183313-5G
ALA183313-1GEA 196.56 USD
ALA183313-1G ALA183313250MG ALA183313-25G ALA183313-5G
6-Acetamidopicolinic acid ≥98%
6-Acetamidopicolinic acid
Formula: C₈H₈N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00233707
CAS Number: 26893-72-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=099af28dbd470f50a41e34cdd7e7ecb60f1d5808&vid=099af28dbd470f50a41e34cdd7e7ecb60f1d5808&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR