H-Deg-OH ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Amino-2-ethylbutanoic acid, H-Deg-OH, 3-Aminopentane-3-carboxylic acid

A183201-1G A183201-250MG A183201-5G
ALA183201-1GEA 47.41 USD
ALA183201-1G ALA183201250MG ALA183201-5G
H-Deg-OH ≥98%
H-Deg-OH
Formula: C₆H₁₃NO₂
MW: 131.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01737173
CAS Number: 2566-29-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=713b1efd55a226dc3e18655de13219883fa22704&vid=713b1efd55a226dc3e18655de13219883fa22704&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR