2-Amino-3-iodo-5-nitropyridine ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
3-Iodo-5-nitropyridin-2-amine, 3-Iodo-5-nitro-2-pyridylamine
A183181-1G
A183181-250MG
A183181-25G
A183181-5G
ALA183181-1GEA
54.06
USD
ALA183181-1G
ALA183181250MG
ALA183181-25G
ALA183181-5G
2-Amino-3-iodo-5-nitropyridine ≥97%
2-Amino-3-iodo-5-nitropyridine
Formula:
C₅H₄IN₃O₂ MW: 265.01 Melting Pt: 231-236 °C |
MDL Number:
MFCD07368863 CAS Number: 25391-56-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...