4'-Amino-2'-hydroxyacetophenone ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1-(4-Amino-2-hydroxyphenyl)ethanone, 1-(4-Amino-2-hydroxyphenyl)ethan-1-one

A183120-1G A183120-5G
ALA183120-1GEA 18.91 USD
ALA183120-1G ALA183120-5G
4'-Amino-2'-hydroxyacetophenone ≥98%
4'-Amino-2'-hydroxyacetophenone
Formula: C₈H₉NO₂
MW: 151.1600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00100636
CAS Number: 2476-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ccec97e0ab3e0c39f2f599346b8be03ab293b8e2&vid=ccec97e0ab3e0c39f2f599346b8be03ab293b8e2&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR