2-Chlorobenzo[d]thiazol-6-amine ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 6-Amino-2-chlorobenzothiazole

A183076-10G A183076-1G A183076-250MG A183076-5G
ALA183076-10GEA 407.46 USD
ALA183076-10G ALA183076-1G ALA183076250MG ALA183076-5G
2-Chlorobenzo[d]thiazol-6-amine ≥97%
2-Chlorobenzo[d]thiazol-6-amine
Formula: C₇H₅ClN₂S
MW: 184.646 g/mol
MDL Number: MFCD08276901
CAS Number: 2406-90-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3310cb3f504d31857baec8ccbccd501c65a29f81&vid=3310cb3f504d31857baec8ccbccd501c65a29f81&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR