4-Amino-3-nitrobenzenesulfonamide ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A183052-1G A183052-250MG A183052-25G A183052-5G
ALA183052-1GEA 116.76 USD
ALA183052-1G ALA183052250MG ALA183052-25G ALA183052-5G
4-Amino-3-nitrobenzenesulfonamide ≥95%
4-Amino-3-nitrobenzenesulphonamide
Formula: C₆H₇N₃O₄S
Storage Temperature: Ambient
MDL Number: MFCD00116612
CAS Number: 2360-19-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=420bd9ad575e94556f55fe7c7ac9fbfbb0a402f4&vid=420bd9ad575e94556f55fe7c7ac9fbfbb0a402f4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR