Ac-Lys(Boc)-OH ≥97%

Supplier: ALADDIN SCIENTIFIC

Synonyms: Nα-Acetyl-Nepsilon-Boc-L-lysine

A183045-100G A183045-1G A183045-25G A183045-5G
ALA183045-100GEA 350.46 USD
ALA183045-100G ALA183045-1G ALA183045-25G ALA183045-5G
Ac-Lys(Boc)-OH ≥97%
Ac-Lys(Boc)-OH
Formula: C₁₃H₂₄N₂O₅
MDL Number: MFCD00236761
CAS Number: 23500-04-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49de658e9b3d29b8a7bf9b86f88908f7c5602eea&vid=49de658e9b3d29b8a7bf9b86f88908f7c5602eea&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR