2-Amino-5-chlorothiophenol ≥96%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A183043-1G A183043-250MG A183043-50MG
ALA183043-1GEA 397.01 USD
ALA183043-1G ALA183043250MG ALA183043-50MG
2-Amino-5-chlorothiophenol ≥96%
2-Amino-5-chlorothiophenol
Formula: C₆H₆ClNS
MW: 159.6 g/mol
Storage Temperature: Ambient
CAS Number: 23474-98-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3579de78fbbd0abf7194ec167e4f9e49c7cff39f&vid=3579de78fbbd0abf7194ec167e4f9e49c7cff39f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR