Doxorubicin ≥97%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: Adriamycin, (7S,9S)-7-[(2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione, Doxil, Hydroxydaunorubicin, Adriablastin, Doxorubicinum, Doxorubicin, Myocet

A183027-25MG A183027-500MG A183027-100MG
ALA183027-25MGEA 164.26 USD
ALA183027-25MG ALA183027500MG ALA183027100MG
Doxorubicin ≥97%, Moligand™
Doxorubicin

Adriamycin is a Topoisomerase II catalytic inhibitor, also is a broad spectrum antibiotic used in the treatment of cancers.

Formula: C₂₇H₂₉NO₁₁
MW: 543.52 g/mol
CAS Number: 23214-92-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc07c0311653b9c20a796a2bcf1cac1fa7cd98b&vid=dcc07c0311653b9c20a796a2bcf1cac1fa7cd98b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR