2-Amino-N-methylacetamide ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A182951-10G A182951-1G A182951-5G
ALA182951-10GEA 347.61 USD
ALA182951-10G ALA182951-1G ALA182951-5G
2-Amino-N-methylacetamide ≥97%
2-Amino-N-methylacetamide
Formula: C₃H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD06375945
CAS Number: 22356-89-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb92cff0f4624a30b1d5ae344b69232a71e14ee0&vid=eb92cff0f4624a30b1d5ae344b69232a71e14ee0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR