2-Amino-3-chloro-5-nitropyridine ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A182945-100MG A182945-1G A182945-250MG A182945-25G A182945-5G
ALA182945100MGEA 10.36 USD
ALA182945100MG ALA182945-1G ALA182945250MG ALA182945-25G ALA182945-5G
2-Amino-3-chloro-5-nitropyridine ≥97%
2-Amino-3-chloro-5-nitropyridine
Formula: C₅H₄ClN₃O₂
MW: 173.5600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00160299
CAS Number: 22353-35-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c67267d2c2c8924c9e4fa99099d740d07c9eecf&vid=6c67267d2c2c8924c9e4fa99099d740d07c9eecf&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR