7-Amino-1-tetralone ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A182867-100MG
A182867-1G
A182867-250MG
A182867-5G
ALA182867100MGEA
14.16
USD
ALA182867100MG
ALA182867-1G
ALA182867250MG
ALA182867-5G
7-Amino-1-tetralone ≥97%
7-Amino-1-tetralone
Formula:
C₁₀H₁₁NO Storage Temperature: Ambient |
MDL Number:
MFCD07369817 CAS Number: 22009-40-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...