2-Aminopyrimidine-4-carboxylic acid ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A182823-100MG A182823-1G A182823-250MG
ALA182823100MGEA 15.11 USD
ALA182823100MG ALA182823-1G ALA182823250MG
2-Aminopyrimidine-4-carboxylic acid ≥97%
2-Aminopyrimidine-4-carboxylic acid
Formula: C₅H₅N₃O₂
MW: 139.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07783936
CAS Number: 2164-65-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bdad2ab1a915d71ff7daa6f46589be3b8b39d837&vid=bdad2ab1a915d71ff7daa6f46589be3b8b39d837&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR