2-Amino-4,6-dimethoxybenzoic acid ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-amino-4,6-dimethoxybenzoic acid

A182812-1G A182812-5G
ALA182812-1GEA 59.76 USD
ALA182812-1G ALA182812-5G
2-Amino-4,6-dimethoxybenzoic acid ≥95%
2-Amino-4,6-dimethoxybenzoic acid
Formula: C₉H₁₁NO₄
MW: 197.1900 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09031989
CAS Number: 21577-57-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91ea869b67a47335867d7f46e7cacf284ed19398&vid=91ea869b67a47335867d7f46e7cacf284ed19398&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR