1-Aminocyclobutanecarboxylic acid methyl ester ≥98%
Supplier: ALADDIN SCIENTIFIC
Warning
|
Synonyms:
1-Amino-cyclobutanecarboxylic acid methyl ester, Methyl 1-aminocyclobutanecarboxylate
A182810-250MG
A182810-5G
A182810-1G
ALA182810250MGEA
19.86
USD
ALA182810250MG
ALA182810-5G
ALA182810-1G
1-Aminocyclobutanecarboxylic acid methyl ester ≥98%
1-Aminocyclobutanecarboxylic acid methyl ester
Formula:
C₆H₁₁NO₂ MW: 129.16 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01318265 CAS Number: 215597-35-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...