2-Amino-4,5-dimethyl-1H-pyrrole-3-carbonitrile ≥97%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
2-Amino-3-cyano-4,5-dimethylpyrrole
A182768-1G
A182768-5G
ALA182768-1GEA
14.16
USD
ALA182768-1G
ALA182768-5G
2-Amino-4,5-dimethyl-1H-pyrrole-3-carbonitrile ≥97%
2-Amino-4,5-dimethyl-1H-pyrrole-3-carbonitrile
Formula:
C₇H₉N₃ MW: 135.17 g/mol |
MDL Number:
MFCD00849263 CAS Number: 21392-51-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...