2-Amino-4,5-dichlorobenzoic acid ≥96%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
A182678-1G
A182678-250MG
A182678-5G
ALA182678-1GEA
60.71
USD
ALA182678-1G
ALA182678250MG
ALA182678-5G
2-Amino-4,5-dichlorobenzoic acid ≥96%
2-Amino-4,5-dichlorobenzoic acid
Formula:
C₇H₅Cl₂NO₂ MW: 206.03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08457795 CAS Number: 20776-61-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...