2-Amino-5-bromo-3-methylbenzoic acid ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A182667-1G A182667-5G
ALA182667-1GEA 28.41 USD
ALA182667-1G ALA182667-5G
2-Amino-5-bromo-3-methylbenzoic acid ≥95%
2-Amino-5-bromo-3-methylbenzoic acid
Formula: C₈H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD05664860
CAS Number: 206548-13-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=246ed3962ea67403701a37beb4051b572705f65c&vid=246ed3962ea67403701a37beb4051b572705f65c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR