4-Bromobenzo[d]thiazol-2-amine ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Amino-4-bromobenzo[d]thiazole

A182605-100G A182605-1G A182605-250MG A182605-25G A182605-5G
ALA182605-100GEA 1847.66 USD
ALA182605-100G ALA182605-1G ALA182605250MG ALA182605-25G ALA182605-5G
4-Bromobenzo[d]thiazol-2-amine ≥98%
4-Bromobenzo[d]thiazol-2-amine
Formula: C₇H₅BrN₂S
MW: 229.097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04971840
CAS Number: 20358-02-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7eb5d4d46a21bbac1ec1147e5a8f8f96765e8a1e&vid=7eb5d4d46a21bbac1ec1147e5a8f8f96765e8a1e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR