Ammonium benzenesulfonate ≥95%

Supplier: ALADDIN SCIENTIFIC

A182444-100G A182444-1G A182444-25G A182444-5G
ALA182444-100GEA 455.91 USD
ALA182444-100G ALA182444-1G ALA182444-25G ALA182444-5G
Ammonium benzenesulfonate ≥95%
Ammonium benzenesulfonate
Formula: C₆H₉NO₃S
Storage Temperature: Ambient
MDL Number: MFCD00191294
CAS Number: 19402-64-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c0655340a64ddefc756889820cbd3f93f3b5184&vid=5c0655340a64ddefc756889820cbd3f93f3b5184&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR