2-Amino-7-bromoquinazoline 96

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 7-Bromoquinazolin-2-amine

A182398-5G A182398-100MG A182398-10G A182398-1G A182398-250MG
ALA182398-5GEA 423.61 USD
ALA182398-5G ALA182398100MG ALA182398-10G ALA182398-1G ALA182398250MG
2-Amino-7-bromoquinazoline 96
2-Amino-7-bromoquinazoline
Formula: C₈H₆BrN₃
MW: 224.0600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09027698
CAS Number: 190274-15-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf2475fa26c1792fb41c68c7f4b2b8e5b9e5eb73&vid=cf2475fa26c1792fb41c68c7f4b2b8e5b9e5eb73&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR