2-Amino-5-methoxybenzamide ≥98%

Supplier: Aladdin Scientific

A182361-1G A182361-250MG A182361-25G A182361-5G
ALA182361-1GEA 27.46 USD
ALA182361-1G ALA182361250MG ALA182361-25G ALA182361-5G
2-Amino-5-methoxybenzamide ≥98%
2-Amino-5-methoxybenzamide
Formula: C₈H₁₀N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD12674808
CAS Number: 1882-71-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3872d823d7eea76dea6e274774d638ff036fa9c5&vid=3872d823d7eea76dea6e274774d638ff036fa9c5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR