6-Amino-5-bromonicotinic acid ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A182227-1G A182227-250MG A182227-5G
ALA182227-1GEA 14.16 USD
ALA182227-1G ALA182227250MG ALA182227-5G
6-Amino-5-bromonicotinic acid ≥97%
6-Amino-5-bromonicotinic acid
Formula: C₆H₅BrN₂O₂
MW: 217.0200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11109515
CAS Number: 180340-69-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c42ab95dde80c79c50b17bcc305e8c57e2aaf600&vid=c42ab95dde80c79c50b17bcc305e8c57e2aaf600&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR