4-Amino-3-methoxybenzonitrile 96

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Cyano-2-methoxy-aniline

A182168-1G A182168-250MG
ALA182168-1GEA 76.86 USD
ALA182168-1G ALA182168250MG
4-Amino-3-methoxybenzonitrile 96
4-Amino-3-methoxybenzonitrile
Formula: C₈H₈N₂O
MW: 148.1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13250177
CAS Number: 177476-76-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cfe128064869ca58e80b37bdec1eb1a11603b87&vid=1cfe128064869ca58e80b37bdec1eb1a11603b87&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR