2-Aminopyrimidine-5-carbonitrile ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION

A182142-1G A182142-5G
ALA182142-1GEA 28.41 USD
ALA182142-1G ALA182142-5G
2-Aminopyrimidine-5-carbonitrile ≥95%
2-Aminopyrimidine-5-carbonitrile
Formula: C₅H₄N₄
MW: 120.1121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00232791
CAS Number: 1753-48-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54098b42818b3277bf7b4bc746e23ccb301f7277&vid=54098b42818b3277bf7b4bc746e23ccb301f7277&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR