2-Amino-4-(3,4-difluorophenyl)thiazole ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-(3,4-Difluorophenyl)thiazol-2-amine, 2-Amino-4-(3,4-difluorophenyl)-1,3-thiazole

A182118-1G A182118-5G
ALA182118-1GEA 64.51 USD
ALA182118-1G ALA182118-5G
2-Amino-4-(3,4-difluorophenyl)thiazole ≥98%
2-Amino-4-(3,4-difluorophenyl)thiazole
Formula: C₉H₆F₂N₂S
MW: 212.218 g/mol
Melting Pt: 121-125 °C
MDL Number: MFCD00052876
CAS Number: 175135-32-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79d417af8f224e5e796b8f9ba05a3bc36bccf67e&vid=79d417af8f224e5e796b8f9ba05a3bc36bccf67e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR