3-Amino-2,4-dichloropyridine ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A182093-250MG A182093-25G A182093-5G
ALA182093250MGEA 9.41 USD
ALA182093250MG ALA182093-25G ALA182093-5G
3-Amino-2,4-dichloropyridine ≥97%
3-Amino-2,4-dichloropyridine
Formula: C5H4Cl2N2
MW: 163.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09750301
CAS Number: 173772-63-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14ccf3ffffc2ea6437c5d6511109c3b1b2133bb2&vid=14ccf3ffffc2ea6437c5d6511109c3b1b2133bb2&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR