3-Amino-6-bromopyrazine-2-carbonitrile ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
A182065-100MG
A182065-10G
A182065-1G
A182065-250MG
A182065-25G
A182065-5G
ALA182065100MGEA
12.26
USD
ALA182065100MG
ALA182065-10G
ALA182065-1G
ALA182065250MG
ALA182065-25G
ALA182065-5G
3-Amino-6-bromopyrazine-2-carbonitrile ≥97%
3-Amino-6-bromopyrazine-2-carbonitrile
Formula:
C₅H₃BrN₄ Storage Temperature: Ambient |
MDL Number:
MFCD06658300 CAS Number: 17231-51-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...