5-Fluoropyrimidin-2-amine ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 2-Amino-5-fluoropyrimidine

A181981-1G A181981-250MG A181981-25G A181981-5G
ALA181981-1GEA 55.01 USD
ALA181981-1G ALA181981250MG ALA181981-25G ALA181981-5G
5-Fluoropyrimidin-2-amine ≥98%
5-Fluoropyrimidin-2-amine
Formula: C4H4FN3
MDL Number: MFCD11846500
CAS Number: 1683-85-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6bab365d8b8e33bb694a6b9e333b320a2fcedc2&vid=e6bab365d8b8e33bb694a6b9e333b320a2fcedc2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR