2-Amino-5-bromobenzamide ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A181899-1G A181899-250MG A181899-5G
ALA181899-1GEA 10.36 USD
ALA181899-1G ALA181899250MG ALA181899-5G
2-Amino-5-bromobenzamide ≥98%
2-Amino-5-bromobenzamide
Formula: C₇H₇BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD04037881
CAS Number: 16313-66-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adaf33ddaba05fa8a96df9ffc8edf599460c8e01&vid=adaf33ddaba05fa8a96df9ffc8edf599460c8e01&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR