4-Acetyl-1H-pyrrole-2-carboxylic acid ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A181870-1G A181870-250MG A181870-5G
ALA181870-1GEA 117.71 USD
ALA181870-1G ALA181870250MG ALA181870-5G
4-Acetyl-1H-pyrrole-2-carboxylic acid ≥95%
4-Acetyl-1H-pyrrole-2-carboxylic acid
Formula: C₇H₇NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD02571849
CAS Number: 16168-93-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8d41e364a60bc000e0b342bfa23963854b8661f&vid=d8d41e364a60bc000e0b342bfa23963854b8661f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR