4-Acetoxycinnamic acid ≥96%

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 3-(4-Acetoxyphenyl)acrylic acid

A181741-25G A181741-5G
ALA181741-25GEA 197.51 USD
ALA181741-25G ALA181741-5G
4-Acetoxycinnamic acid ≥96%
4-Acetoxycinnamic acid
Formula: C₁₁H₁₀O₄
MW: 206.2 g/mol
Melting Pt: 198-210°C
MDL Number: MFCD00016847
CAS Number: 15486-19-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4af379a5b575dd403566282df9d7934a613f68cc&vid=4af379a5b575dd403566282df9d7934a613f68cc&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR