2-Amino-6-fluorobenzaldehyde ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A181670-100MG A181670-1G A181670-250MG A181670-5G
ALA181670100MGEA 58.81 USD
ALA181670100MG ALA181670-1G ALA181670250MG ALA181670-5G
2-Amino-6-fluorobenzaldehyde ≥98%
2-Amino-6-fluorobenzaldehyde
Formula: C₇H₆FNO
Storage Temperature: Refrigerator
MDL Number: MFCD12400881
CAS Number: 151585-93-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e094a084964931fe5c5b735893e4d5d091cc677&vid=0e094a084964931fe5c5b735893e4d5d091cc677&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR