2-Amino-3-methoxybenzonitrile 96

Supplier: ALADDIN SCIENTIFIC
Warning

A181607-100MG A181607-10G A181607-1G A181607-250MG A181607-25G A181607-5G
ALA181607100MGEA 19.86 USD
ALA181607100MG ALA181607-10G ALA181607-1G ALA181607250MG ALA181607-25G ALA181607-5G
2-Amino-3-methoxybenzonitrile 96
2-Amino-3-methoxybenzonitrile
Formula: C₈H₈N₂O
MW: 148.1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09744022
CAS Number: 148932-68-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c20724457dfd661fb5f169f6c6b9d30532fcb1e&vid=0c20724457dfd661fb5f169f6c6b9d30532fcb1e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR